Stage Micrometer Model SM-12, Electron Microscopy Sciences
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
: The Opal and Quartz are offered as a special only and must be requested.
: All EMS micrometers come in a sturdy wooden box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.