Stage Micrometer Model SM-4, Electron Microscopy Sciences
About this item
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
: The Opal and Quartz are offered as a special only and must be requested.
: All EMS micrometers come in a sturdy wooden box.
2 Options Available Below
Product Details & Documents
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
: The Opal and Quartz are offered as a special only and must be requested.
: All EMS micrometers come in a sturdy wooden box.
Product Family Options
Product Information
- AvailabilityPrice
- 102093-118Stage Micrometer SM-4 Chrome on Glass with CalibrationSupplier Number: 60210-4PGC
- 102680-930Stage Micrometer SM-4, Chrome on glassSupplier Number: 60210-4PG