ContactUsLinkComponent
TEM Grid Holder on Pin, Electron Microscopy Sciences
TEM Grid Holder on Pin, Electron Microscopy Sciences
Catalog # 102098-298
TEM Grid Holder on Pin, Electron Microscopy Sciences
Catalog # 102098-298
Supplier Number:  75949-03
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    TEM grid holder on pin
  • Cat. No.:
    102098-298
  • Supplier no.:
    75949-03
Specifications
About this item
This EMS new release allows for the holding of up to 4 grids. The grid locations are all numbered.
  • Made from aluminum with a brass screw
  • This holder allows to image and analyze specimens on TEM grids in the SEM
The overall diameter of the holder is 1" (25 mm) with a 1/8" Pin (3.2 mm) and a longer pin 0.6" (15 mm).