About this item
These PTFE molds allow for easy removal of 10 plastic blocks when using the Histobloc® system.
- Material-saving volume
- Easy-to-cut shape
- Unlimited shelf life
- Easy removal of the embedded specimens because of the conical shape and the Teflon® material
- Excellent temperature dissipation by way of refined steel bottom
Three types types are available: the Histoform Q and the Histoform S and the Histoform N. Accurate and easy fixing of the specimen blocks byspecial grooves were achieved with Histoform S, in cooperation with renowned users.
The mold cavities measure 10W×16H×6.5D mm in 'S' and 20W×16H×10D mm in 'Q' and in 'N' 9W×18H×5D mm.