Specifications
- Description:Grid Transporting Box
- Cat. No.:102097-450
- Supplier no.:71173-01
Specifications
About this item
This unique Box allows for the storage and transportation of grids without any worry of them moving around or being damaged.
The Plate is made from Silicone and has 54 individual compartments 10 mm in size. Each compartment can hold 3 grids and the grids can be easily picked up from the Silicone surface.