Orders LinkContactUsLinkComponent
Disposable Micro Tissue Grinder, Electron Microscopy Sciences
Disposable Micro Tissue Grinder, Electron Microscopy Sciences
Disposable Micro Tissue, 0.75 ml
Catalog # 102095-708
Supplier #:  64803-07
Disposable Micro Tissue Grinder, Electron Microscopy Sciences
Catalog # 102095-708
Disposable Micro Tissue Grinder, Electron Microscopy Sciences
Disposable Micro Tissue Grinder, Electron Microscopy Sciences
Disposable Micro Tissue, 0.75 ml
Price
$463.55
Log in for availability
Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The disposable micro tissue grinder allows for homogenized tissue micro samples without cross contamination. This unit is especially useful for radioactive samples.
  • Eliminates cross contamination
  • Useful for radioactive samples
  • Small grinder area for high sample recovery
  • Made from Wheaton 33 low extractable borosilicate glas
  • Autocalvable
No washing is needed; just disposed the set after use. Mortar dimensions are 10 x 44 mm. Pestle size is 7 mm sphere.
Specifications
  • Description:
    Disposable Micro Tissue
  • Size:
    0.75 ml