Orders LinkContactUsLinkComponent
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Certificates
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Similar to the all glass Potter-Elvehjem style, but micro size. Ideal for extremely precise work.
  • Made from Wheaton 33 low extractable borosilicate glass
  • Micro size for extremely precise work
  • Reservoir and pouring lip are incorporated into the design
  • Autoclavable
This unit is designed for delicate hand operation. Clearance between pestle and tube is 0.004” to 0.006” (0.1 – 015 mm).
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 102095-612
    Potter-Elvehjem Micro Tissue Grinder
    Potter-Elvehjem Micro Tissue Grinder
    0.1 ml
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $287.46
    $287.46
    Pack of 2
       $287.46
    Supplier Number: 64794-01
  • 102095-614
    Potter-Elvehjem Micro Tissue Grinder Pestle only
    Potter-Elvehjem Micro Tissue Grinder Pestle only
    0.1 ml
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $247.33
    $247.33
    Pack of 2
       $247.33
    Supplier Number: 64794-70
  • 102095-616
    Potter-Elvehjem Micro Tissue Grinder Mortar only
    Potter-Elvehjem Micro Tissue Grinder Mortar only
    0.1 ml
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $166.77
    $166.77
    Pack of 2
       $166.77
    Supplier Number: 64794-80