Orders LinkContactUsLinkComponent
Ti disc Embedding Ring, Electron Microscopy Sciences
Ti disc Embedding Ring, Electron Microscopy Sciences
TI Embedding Rings
Catalog # 102092-804
Supplier #:  55110-TIR
Ti disc Embedding Ring, Electron Microscopy Sciences
Catalog # 102092-804
Ti disc Embedding Ring, Electron Microscopy Sciences
Ti disc Embedding Ring, Electron Microscopy Sciences
TI Embedding Rings
Price
$179.93
Log in for availability
Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ti disc Embedding Ring for TEM sample preparation. Used for the most difficult step of cross-sectional thinning is the mechanical preparation of the sample, cutting a cross section and then embedding it into an appropriately designed ring.
Using this ring, embedding cross sections is greatly simplified, since the samples do not need to be glued together prior to embedding. In order to fix the samples in the ring it is deformed mechanically by an appropriate tool at the two points marked by the arrows. This deformation will both hold the sample in the ring and press both sample pieces together before gluing. Ti disc is also offered for embedding bulk samples of brittle material for plan-view TEM investigations.
Delivery information: The rings supplied in 5 sizes 0.6, 0.8, 1.0, 1.2, and 1.3 mm.
Specifications
  • Description:
    TI Embedding Rings