Orders LinkContactUsLinkComponent
Standard Reticles with Indexed Grid Scales, Electron Microscopy Sciences
Standard Reticles with Indexed Grid Scales, Electron Microscopy Sciences
Standard Reticles with Indexed Grid Scales, Electron Microscopy Sciences
Certificates
Standard Reticles with Indexed Grid Scales, Electron Microscopy Sciences
Standard Reticles with Indexed Grid Scales, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • RET 83
    5×5×1.0 mm with 0.01 mm line
    Log in for availability
    Log in for availability
    Each
    Each - $320.01
    $320.01
    Each
       $320.01
    Supplier Number: 60286-19
  • RET 83
    5×5×1.0 mm with 0.01 mm line
    Log in for availability
    Log in for availability
    Each
    Each - $320.01
    $320.01
    Each
       $320.01
    Supplier Number: 60286-21
  • RET 80
    10×10×0.5 mm with 0.025 mm line
    Log in for availability
    Log in for availability
    Each
    Each - $320.01
    $320.01
    Each
       $320.01
    Supplier Number: 60287-19
  • RET 80
    10×10×0.5 mm with 0.025 mm line
    Log in for availability
    Log in for availability
    Each
    Each - $320.01
    $320.01
    Each
       $320.01
    Supplier Number: 60287-21