Order Entry
United States
Orders LinkContactUsLinkComponent
Dissecting Kit for Botany, Electron Microscopy Sciences

Dissecting Kit for Botany, Electron Microscopy Sciences

Dissecting Kit for Botany
Catalog # 102098-096
Supplier #:  72951-20
Dissecting Kit for Botany, Electron Microscopy Sciences
Catalog # 102098-096
Dissecting Kit for Botany, Electron Microscopy Sciences
Dissecting Kit for Botany, Electron Microscopy Sciences

Dissecting Kit for Botany

Price
$101.18
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This botany dissection kit includes the following:

  • Dissecting teasing needle angular with metal chuck brass-chromed
  • Dissecting teasing needle straight with metal chuck brass-chromed
  • Ruler 6”
  • Dissecting forceps 4.5” curved fine point with guide-pin
  • Dissecting forceps medium point 4.5” with guide-pin
  • Dissecting Iris scissors 4.5”
  • Dissecting scalpel handle #3
  • No. 10 scalpel blade, 10 pcs
  • Glass dropper
  • Replacement dissecting needle straight, 6/pk
  • Replacement dissecting needle angular, 6/pk
  • Vinyl storage, zippered case with liner.
Specifications
  • Description:
    Dissecting Kit for Botany

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.