Orders LinkContactUsLinkComponent
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Certificates
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Paraffin Tissue Microarrays, Electron Microscopy Sciences
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Paraffin tissue microarrays (TMAs) are becoming more common in the laboratories. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process. Arraymold features include:
  • Allow pathologist or researcher to view many different samples on the same single slide
  • Helpful in finding IHC markers for control or quality assurance from hundreds of samples
  • Staining markers will be more consistent when hundreds of samples are on one slide
  • Used less costly antibody
  • Small enough to be stored in a drawer
  • Rubber arraymold can be reused many times without losing its flexibility
Current techniques including organizing, punching, and inserting tissues is time consuming and involving costly equipments.This Arraymold is the simple tool but great answer for this current technique for laboratories to perform TMAs with less time consuming, excellent results. With this Arraymold, you can process up to 150 specimens onto one single slide with very little time consuming, minimal technical training.
Product Family Options
Product Information
  • Includes
    Availability
    Price
  • 102096-518
    Arraymold Kit A, 60 Cores
    Arraymold Kit A, 60 Cores
    10 Dermal Needles (2 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (2 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,648.24
    $1,648.24
    Each
       $1,648.24
    Supplier Number: 69130-10
  • 102096-520
    Arraymold Kit B, 150 Cores
    Arraymold Kit B, 150 Cores
    10 Dermal Needles (1.5 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (1.5 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,648.24
    $1,648.24
    Each
       $1,648.24
    Supplier Number: 69130-20
  • 102096-522
    Arraymold Kit C, 15 Cores
    Arraymold Kit C, 15 Cores
    10 Dermal Needles (4 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (4 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,648.24
    $1,648.24
    Each
       $1,648.24
    Supplier Number: 69130-40
  • 101765-094
    ArrayMold D 36 Core, 2 mm
    ArrayMold D 36 Core, 2 mm
    10 Dermal Needles (2 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (2 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,116.77
    $1,116.77
    Each
       $1,116.77
    Supplier Number: 69130-15
  • 101765-096
    ArrayMold E 72 Core, 1.5 mm
    ArrayMold E 72 Core, 1.5 mm
    10 Dermal Needles (1.5 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (1.5 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,116.77
    $1,116.77
    Each
       $1,116.77
    Supplier Number: 69130-25
  • 101765-098
    ArrayMold F 35 Core, 3 mm
    ArrayMold F 35 Core, 3 mm
    10 Dermal Needles (3 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (4 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,648.24
    $1,648.24
    Each
       $1,648.24
    Supplier Number: 69130-45
  • 101765-100
    ArrayMold G 24 Core, 3.5 mm
    ArrayMold G 24 Core, 3.5 mm
    10 Dermal Needles (3.5 mm), 2 Stylets, 1 Brass Plate, 1 Arraymold (4 mm), 1 DVD
    Log in for availability
    Log in for availability
    Each
    Each - $1,122.49
    $1,122.49
    Each
       $1,122.49
    Supplier Number: 69130-55
  • 102096-524
    Arraymold package Kit 1
    Arraymold package Kit 1
    Arraymold Kits A & B
    Log in for availability
    Log in for availability
    Each
    Each - $3,294.76
    $3,294.76
    Each
       $3,294.76
    Supplier Number: 69131-01
  • 102096-526
    Arraymold package Kit 2
    Arraymold package Kit 2
    Arraymold Kits A & C
    Log in for availability
    Log in for availability
    Each
    Each - $3,294.76
    $3,294.76
    Each
       $3,294.76
    Supplier Number: 69131-02
  • 102096-528
    Arraymold package Kit 3
    Arraymold package Kit 3
    Arraymold Kits B & C
    Log in for availability
    Log in for availability
    Each
    Each - $3,294.76
    $3,294.76
    Each
       $3,294.76
    Supplier Number: 69131-03
  • 102096-530
    Arraymold package Kit 4
    Arraymold package Kit 4
    Arraymold Kits A, B & C
    Log in for availability
    Log in for availability
    Each
    Each - $4,906.97
    $4,906.97
    Each
       $4,906.97
    Supplier Number: 69131-04
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Styler 4.0 mm
    Styler 4.0 mm
    101764-970
    Log in for availability
    Each - $64.50
Product Information
  • Catalog #
    Availability
    Price
  • Styler 3.5 mm
    Styler 3.5 mm
    101764-968
    Log in for availability
    Each - $64.50
Product Information
  • Catalog #
    Availability
    Price
  • Styler 3.0 mm
    Styler 3.0 mm
    101764-966
    Log in for availability
    Each - $64.50
Product Information
  • Catalog #
    Availability
    Price
  • Styler 1.5 mm
    Styler 1.5 mm
    101764-962
    Log in for availability
    Each - $64.50