About this item
This butyl acetate and acrylic resin mixture is the first mounting medium developed for coverslipping directly from d-limonene based clearants. Superior results are also with xylene. CitraMount® medium is recommended for manual coversliping only as it may cause damage to the valves of automated coverslippers.
- Clear seal with drying time is less than 24 hours
- efractive index is 1.42
- Viscosity is 90 cPs
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- UN No.:1123
- Density:4 g/cm³ (33.38 lbs/gal)
- Boiling Point:110 °C (230 °F)