About this item
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes.
- Thermostatic heat control
- Anodized black surface provides contrast
- LED temperature display
- Thermostat setting range from room temperature to 70 °C