ContactUsLinkComponent
Simport MEGA Double Cytology Funnels, Electron Microscopy Sciences
Simport MEGA Double Cytology Funnels, Electron Microscopy Sciences
  102094-450
undefined
Simport MEGA Double Cytology Funnels, Electron Microscopy Sciences
  102094-450
 :  62754-10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Single Funnel with Cap
  • Cat. No.:
    102094-450
  • Supplier no.:
    62754-10
This ALL PLASTIC CytoSep™ MEGA Funnel provides an easy, efficient and cost effective method of producing high quality thin-layer slide preparations. It eliminates the needs for stainless steel clips. It is ideally for preparation of large volume samples. The larger funnel is quick and easy to remove. The large rectangular cell deposition area of 22 x 14.75 mm (325 mm squared) provides for up to 12 times the sample volume (6 ml) of the single Cyto™ Cytology Funnel. Strategically placed baffles inhibit cell setting which results in uniform cell deposition and excellent quality slide preparations. This MEGA funnels and cap minimize user exposure to pathogens while reducing the risk of specimen cross contamination. Can be used wtih both air dried and fixed preparations.