Spray droplet sizing reticle for size and distribution assessments of aersol droplets. Used in conjunction with a 40x Microscope for direct measurements of droplets from 50 to 400 μm diameter.
The actual pattern sizes are 50, 100, 200, and 400 μm.