Orders LinkContactUsLinkComponent
Indexed Grids, Electron Microscopy Sciences
Indexed Grids, Electron Microscopy Sciences
Certificates
Indexed Grids, Electron Microscopy Sciences
Indexed Grids, Electron Microscopy Sciences
20 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful for particle counting and in area of specimen determinations.
  • NE10A: numbered grid 5×5 mm, 0.5 mm pitch
  • NE11A: numbered grid 10×10 mm, 1.0 mm pitch
  • NE34A: numbered grid 1×1 mm, 0.1 mm pitch
  • NE71: index pattern 20×20 grid of 0.5 mm squares
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100499-146
    Graticle NE10A
    Graticle NE10A
    16 mm
    Log in for availability
    Log in for availability
    Each
    Each - $227.91
    $227.91
    Each
       $227.91
    Supplier Number: 68011-16A
  • 100499-176
    Graticle NE11A
    Graticle NE11A
    16 mm
    Log in for availability
    Log in for availability
    Each
    Each - $183.63
    $183.63
    Each
       $183.63
    Supplier Number: 68016-16
  • 100499-190
    Graticle NE34A
    Graticle NE34A
    16 mm
    Log in for availability
    Log in for availability
    Each
    Each - $244.73
    $244.73
    Each
       $244.73
    Supplier Number: 68018-16A
  • 100499-178
    Graticle NE11A
    Graticle NE11A
    19 mm
    Log in for availability
    Log in for availability
    Each
    Each - $213.43
    $213.43
    Each
       $213.43
    Supplier Number: 68016-19
  • 100499-194
    Graticle NE34A
    Graticle NE34A
    19 mm
    Log in for availability
    Log in for availability
    Each
    Each - $244.73
    $244.73
    Each
       $244.73
    Supplier Number: 68018-19A
  • 102096-110
    Graticle NE10A
    Graticle NE10A
    19 mm
    Log in for availability
    Log in for availability
    Each
    Each - $251.89
    $251.89
    Each
       $251.89
    Supplier Number: 68011-19A
  • 103303-602
    Graticle NE11A
    Graticle NE11A
    20.4 mm
    Log in for availability
    Log in for availability
    Each
    Each - $188.04
    $188.04
    Each
       $188.04
    Supplier Number: 68016-20.4
  • 102096-050
    Graticle NE71
    Graticle NE71
    21 mm
    Log in for availability
    Log in for availability
    Each
    Each - $211.27
    $211.27
    Each
       $211.27
    Supplier Number: 67981-21
  • 100499-198
    Graticle NE34A
    Graticle NE34A
    21 mm
    Log in for availability
    Log in for availability
    Each
    Each - $281.00
    $281.00
    Each
       $281.00
    Supplier Number: 68018-21A
  • 100499-180
    Graticle NE11A
    Graticle NE11A
    21 mm
    Log in for availability
    Log in for availability
    Each
    Each - $181.65
    $181.65
    Each
       $181.65
    Supplier Number: 68016-21
  • 102096-052
    Graticle NE71
    Graticle NE71
    23 mm
    Log in for availability
    Log in for availability
    Each
    Each - $200.19
    $200.19
    Each
       $200.19
    Supplier Number: 67981-23
  • 103303-604
    Graticle NE11A
    Graticle NE11A
    23 mm
    Log in for availability
    Log in for availability
    Each
    Each - $188.04
    $188.04
    Each
       $188.04
    Supplier Number: 68016-23
  • 102096-054
    Graticle NE71
    Graticle NE71
    24 mm
    Log in for availability
    Log in for availability
    Each
    Each - $296.54
    $296.54
    Each
       $296.54
    Supplier Number: 67981-24
  • 103303-606
    Graticle NE11A
    Graticle NE11A
    24.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $228.89
    $228.89
    Each
       $228.89
    Supplier Number: 68016-24.5
  • 103303-608
    Graticle NE11A
    Graticle NE11A
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $228.89
    $228.89
    Each
       $228.89
    Supplier Number: 68016-25
  • 102096-056
    Graticle NE71
    Graticle NE71
    26 mm
    Log in for availability
    Log in for availability
    Each
    Each - $296.54
    $296.54
    Each
       $296.54
    Supplier Number: 67981-26
  • 103303-610
    Graticle NE11A
    Graticle NE11A
    26 mm
    Log in for availability
    Log in for availability
    Each
    Each - $244.59
    $244.59
    Each
       $244.59
    Supplier Number: 68016-26
  • 103303-612
    Graticle NE11A
    Graticle NE11A
    27 mm
    Log in for availability
    Log in for availability
    Each
    Each - $228.89
    $228.89
    Each
       $228.89
    Supplier Number: 68016-27
  • 102096-058
    Graticle NE71
    Graticle NE71
    Custom
    Log in for availability
    Log in for availability
    Each
    Each - $379.05
    $379.05
    Each
       $379.05
    Supplier Number: 67981-50
  • 102096-112
    Graticle NE10A
    Graticle NE10A
    21 mm
    Log in for availability
    Log in for availability
    Each
    Each - $251.89
    $251.89
    Each
       $251.89
    Supplier Number: 68011-21A