Orders LinkContactUsLinkComponent
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System, Electron Microscopy Sciences
The Matrix system has a simply alpha-numeric identification system. The unit is not solvent or chemically resistant to acids so all stains should be aqueous based only.
  • Stains up to 25 grids at one time or as little as one
The system requires very little stain and you may use different vessels for each stain.
 : Each system includes the Matrix Body and 2 each of the Staining Vessels (red and blue).
Product Family Options
  • Description
  • Grid Staining Matrix System Kit
    Each
    Each - $220.64
    $220.64
    Each
       $220.64
     : 71179-01
  • Matrix Body with Handle and Cover
    Each
    Each - $103.78
    $103.78
    Each
       $103.78
     : 71179-05
  • Staining Vessels, 1 Red and 1 Blue
    Pack of 2
    Pack of 2 - $131.88
    $131.88
    Pack of 2
       $131.88
     : 71179-06
  • Staining Vessel, Blue
    Each
    Each - $69.89
    $69.89
    Each
       $69.89
     : 71179-07
  • Staining Vessel, Red
    Each
    Each - $69.89
    $69.89
    Each
       $69.89
     : 71179-08