Ideally for morphological calibration where two dimensional shapes have to be measured precisely.
Standard includes a set of eight test plates of various shapes, positions, angles and sizes. It also includes a linear measurement scale for calibrating your imaging system. Overall size: 25 x 75 mm (1 x 3”). Positive image / Chrome on glass (PG).