Specialty Labels, Electron Microscopy Sciences
About this item
Preprinted Tough-Spots make labeling and identifying samples quick and easy.
- Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
1 Options Available Below
Product Details & Documents
Preprinted Tough-Spots make labeling and identifying samples quick and easy.
- Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Product Family Options
Product Information
- LegendColorDiameterAvailabilityPrice
- 102098-690Tough-SpotsSupplier Number: 77593-14
- 102098-698Tough-SpotsTough-SpotsRadioactive—3/8"Discontinued-Supplier Number: -
- 102098-684Tough-SpotsTough-SpotsBullseyeDark green7/16"Discontinued-Supplier Number: -
- 102098-686Tough-SpotsTough-SpotsBullseyeCopper7/16"Discontinued-Supplier Number: 77593-12
- 102098-688Tough-SpotsTough-SpotsBullseyeDark purple7/16"Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -