Orders LinkContactUsLinkComponent
Cell Culture Inserts, Electron Microscopy Sciences
Cell Culture Inserts, Electron Microscopy Sciences
Cell Culture Inserts, Electron Microscopy Sciences
Certificates
Cell Culture Inserts, Electron Microscopy Sciences
Cell Culture Inserts, Electron Microscopy Sciences
12 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cell culture inserts with PC membranes.
Polycarbonate Membranes:
Easy cultivation of most cell types without matrix coating
Treated for cell culture
Inserts with 0.02 µm membrane have maximum clarity for microscopy, and are non-auto-fluorescent and highly porous
Polycarbonate membranes in larger pore sizes are adaptable to many uses in cell culture, including transport studies, toxicity tests, chemotaxis studies and electron microscopy
Both membrane types are transparent when wet and suitable for phase contrast and Nomarski Optic Systems
Polycarbonate Membrane Inserts Pre-packed in Multidishes:
For multidishes 6, 12, 24
Treated and tested for cell attachment
Easy cultivation of most cell types without matrix coating
Used in a varity of applications, including transport studies, toxicity tests, chemotaxis studies and electron microscopy
Non-toxic
Packed in sterile Nunclon™ Δ treated multidishes
Packed in resealable bags
Non-pyrogenic
Tabs for better grip/handling
Applications for the TCI include Epithelial polarization studies, Chemotaxix studies, Co-culture experiments, transport studies, electron microscopy, toxicity testing, and modeling of in-vivo barriers.
Suggested working volume, mL is in addition to normal well working volume.
Product Family Options
Product Information
  • Working volume
    Culture area
    Pore size
    Sterility
    Availability
    Price
  • 102097-890
    Cell culture inserts with PC membranes for Multidish 24
    Cell culture inserts with PC membranes for Multidish 24
    0.5 ml
    0.47 cm²
    0.4 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $810.79
    $810.79
    Case of 48
       $810.79
    Supplier Number: 72296-16
  • 102097-892
    Cell culture inserts with PC membranes for Multidish 24
    Cell culture inserts with PC membranes for Multidish 24
    0.5 ml
    0.47 cm²
    3.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $790.14
    $790.14
    Case of 48
       $790.14
    Supplier Number: 72296-17
  • 102097-894
    Cell culture inserts with PC membranes for Multidish 24
    Cell culture inserts with PC membranes for Multidish 24
    0.5 ml
    0.47 cm²
    8.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $809.16
    $809.16
    Case of 48
       $809.16
    Supplier Number: 72296-18
  • 102097-896
    Cell culture inserts with PC membranes for Multidish 12
    Cell culture inserts with PC membranes for Multidish 12
    1.1 ml
    1.13 cm²
    0.4 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $842.13
    $842.13
    Case of 48
       $842.13
    Supplier Number: 72296-19
  • 102097-898
    Cell culture inserts with PC membranes for Multidish 12
    Cell culture inserts with PC membranes for Multidish 12
    1.1 ml
    1.13 cm²
    3.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $842.13
    $842.13
    Case of 48
       $842.13
    Supplier Number: 72296-20
  • 102097-900
    Cell culture inserts with PC membranes for Multidish 12
    Cell culture inserts with PC membranes for Multidish 12
    1.1 ml
    1.13 cm²
    8.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $842.48
    $842.48
    Case of 48
       $842.48
    Supplier Number: 72296-21
  • 102097-902
    Cell culture inserts with PC membranes for Multidish 6
    Cell culture inserts with PC membranes for Multidish 6
    1.5 ml
    3.14 cm²
    0.4 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $446.17
    $446.17
    Case of 48
       $446.17
    Supplier Number: 72296-22
  • 102097-904
    Cell culture inserts with PC membranes for Multidish 6
    Cell culture inserts with PC membranes for Multidish 6
    1.5 ml
    3.14 cm²
    3.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $462.61
    $462.61
    Case of 48
       $462.61
    Supplier Number: 72296-23
  • 102097-906
    Cell culture inserts with PC membranes for Multidish 6
    Cell culture inserts with PC membranes for Multidish 6
    1.5 ml
    3.14 cm²
    8.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $466.68
    $466.68
    Case of 48
       $466.68
    Supplier Number: 72296-24
  • 102097-908
    Cell culture inserts with PC membranes for Multidish 6
    Cell culture inserts with PC membranes for Multidish 6
    1.75 ml
    4.1 cm²
    0.4 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $446.17
    $446.17
    Case of 48
       $446.17
    Supplier Number: 72296-25
  • 102097-910
    Cell culture inserts with PC membranes for Multidish 6
    Cell culture inserts with PC membranes for Multidish 6
    1.75 ml
    4.1 cm²
    3.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $440.39
    $440.39
    Case of 48
       $440.39
    Supplier Number: 72296-26
  • 102097-912
    Cell culture inserts with PC membranes for Multidish 6
    Cell culture inserts with PC membranes for Multidish 6
    1.75 ml
    4.1 cm²
    8.0 µm
    Sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $453.80
    $453.80
    Case of 48
       $453.80
    Supplier Number: 72296-27