Order Entry
United States
ContactUsLinkComponent
Sample Stub Vacuum Desiccators , Electron Microscopy Sciences
Sample Stub Vacuum Desiccators , Electron Microscopy Sciences
Catalog # 101454-046
Minority or Woman-Owned Business Enterprise
undefined
Sample Stub Vacuum Desiccators , Electron Microscopy Sciences
Catalog # 101454-046
Supplier Number:  76550
Minority or Woman-Owned Business Enterprise

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    SEM Stub Vacuum Desiccator
  • Cat. No.:
    101454-046
  • W×D×H:
    13 × 13 × 4.5 mm
  • Chamber dimensions:
    10.4 × 18mm
  • Supplier no.:
    76550

Specifications

About this item

Precision machined from a block of aluminum – anodized black - this Sample Stub Vacuum Desiccators accommodates 18 SEM Mounted Sample Stubs, pin style, under vacuum for year long

13 x 13 x 4.5 cm. Chamber cavity is 10.4 cm diameter and 18 mm deep. Lid is made with 10 mm thick clear acrylic for easyviewing the content. Prevents oxidation from occurring  to the sample during long term storage.