ContactUsLinkComponent
Universal Cassette and Cyclone Holder, Electron Microscopy Sciences
Universal Cassette and Cyclone Holder, Electron Microscopy Sciences
Catalog # 102681-130
undefined
Universal Cassette and Cyclone Holder, Electron Microscopy Sciences
Catalog # 102681-130
Supplier Number:  65230-01
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    25 mm Asbestos TEM Cassette
  • Cat. No.:
    102681-130
  • Supplier no.:
    65230-01
Specifications
About this item
This Universal Cassette and Cyclone Holder allows cassettes and cyclones to be attached onto a workers collar.
Compatible with 37 mm and 25 mm cassettes, and aluminum cyclone manufactured by Zefon and SKC. Weighs only 73 g or 2.5 oz. Comes with 3' of tubing attached.