ContactUsLinkComponent
Epo-Tek® H20S Adhesive, Electron Microscopy Sciences
Epo-Tek® H20S Adhesive, Electron Microscopy Sciences
Catalog # 102091-812
CAS Number:  
undefined
Epo-Tek® H20S Adhesive, Electron Microscopy Sciences
Catalog # 102091-812
Supplier Number:  12672-20S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    H20S  Adhesive
  • Size:
    1 oz
  • Environmentally Preferable:
  • Cat. No.:
    102091-812
  • Supplier no.:
    12672-20S
Specifications
About this item
Epo-Tek® H20S is a modified version of Epo-Tek®H20E
  • Highly reliable
  • Smooth consistency
  • High electrical conductivity
This modified version offers high electrical conductivity, short curing cycles, proven reliability, and the convenient mix ratio, Epo-Tek® H20S is extremely simple to use. Epo-Tek® H20S pot life is 2.5 days and shelf life is one year when store at room temperature.
Epo-Tek® H20S is a highly reliable, two component, silver-filled epoxy with a smooth, thixotropic consistency (mixing ratio 1:1).