Orders LinkContactUsLinkComponent
Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Copper Straight End Effector Type 3
Catalog # 102681-338
Supplier #:  75981-CU
Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Catalog # 102681-338
Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Copper Straight End Effector Type 3
Price
$1,350.00
Log in for availability
Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Type 2 Curved End Effector is designed for low attack angle manipulation and lift-out. The shank is curved to permit low angle approach and solid attachment beneath the sample plane without collision with the surrounding matrix. The Straight End Effector for Xtreme Access probe shaft.
Ordering information: Available in Copper (Cu) Curved End Effector. Available in two versions: Copper (Cu) and Molybdenum (Mo).
Specifications
  • Description:
    Copper Straight End Effector Type 3