ContactUsLinkComponent
Standard Pin Stub SEM Mount Small Non-Slotted Head, Electron Microscopy Sciences
Standard Pin Stub SEM Mount Small Non-Slotted Head, Electron Microscopy Sciences
Catalog # 102091-298
undefined
Standard Pin Stub SEM Mount Small Non-Slotted Head, Electron Microscopy Sciences
Catalog # 102091-298
Supplier Number:  75186
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Pin Stub Non-Slotted Head
  • Material:
    Aluminum
  • Cat. No.:
    102091-298
  • Head diameter:
    25.4 mm
  • Supplier no.:
    75186
  • Pin size:
    8 H×3.1 Ø mm
Specifications
About this item
Non-Slotted head pin stub mounts are for use with Cambridge, Leo, Philips 500, Cameca SEMs. Made from lathe finished ultra-pure aluminium.