Orders LinkContactUsLinkComponent
ProScope HR Microscopes, Electron Microscopy Sciences
ProScope HR Microscopes, Electron Microscopy Sciences
ProScope HR Microscopes, Electron Microscopy Sciences
Certificates
ProScope HR Microscopes, Electron Microscopy Sciences
ProScope HR Microscopes, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The ProScope HR has been a mainstay in science education, law enforcement, and manufacturing quality control worldwide. The software for Windows and Mac will capture stills, video and time lapse. Lenses range 0 to 10×, 30×, 50×, 100×, 200×, 400× and 1000×. The ProScope HR can be connected to analog microscope, telescope or borescope.
  • Accurate, repeatable measurement within 10 µm
  • Modular lenses range from 1× to 1000×
  • No driver needed, just plug and play
  • Lead-free
  • Attaches to other optics like Borescopes and analog microscopes
  • 30-day no questions asked return policy
  • Two year warranty
The ProScope HR is USB Video Class and the ProScope HR can be used with programs like Skype or iChat to show others what this ProScope HR is seeing in real time over the internet.
The new ProScope HR2 features the new state-of-the-art, Aptina imager which was originally made for surveillance, with large pixels that take in more light than other imagers on the market. The Bodelin engineering team customized this imager to allow extensive imaging adjustment and LED intensity control. The most impressive feature is the ability to actually stream true pixels at higher resolutions.
The ProScope HR2 is the only digital microscope in the world that can do this. Other imagers stream at 640×480 and only interpolate to simulate higher resolutions. The total effect in the ProScope HR2 is live video that appears to be 4 to 5 megapixels in clarity while only using an actual 2 megapixel imager. The new cobalt black handle says this is a serious tool yet the price point is equal to the previous ProScope HR, which will remain in the product line at a lower cost. The ProScope products are currently used in tens of thousands of schools, universities, law enforcement forensic labs, manufacturing quality control, medical and tele-medicine.
Certifications: RoHS compliant.
Delivery information: Software for Windows and Mac is included.
Product Family Options
Product Information
  • Availability
    Price
  • 102096-240
    ProScope HR Lab
    ProScope HR Lab
    Log in for availability
    Log in for availability
    Each
    Each - $1,459.40
    $1,459.40
    Each
       $1,459.40
    Supplier Number: 68350-62
  • 102096-242
    ProScope HR2 Lab
    ProScope HR2 Lab
    Discontinued
    Supplier Number: -
  • 102096-244
    ProScope Wireless Lab
    ProScope Wireless Lab
    Discontinued
    Supplier Number: -
  • 102096-246
    ProScope HR Advanced Lab
    ProScope HR Advanced Lab
    Discontinued
    Supplier Number: -
  • 102096-248
    ProScope HR2 Advanced Lab
    ProScope HR2 Advanced Lab
    Discontinued
    Supplier Number: -
  • 102096-250
    ProScope Wireless Advanced Lab
    ProScope Wireless Advanced Lab
    Discontinued
    Supplier Number: -
  • 102096-228
    ProScope HR CSI Science Level 1
    ProScope HR CSI Science Level 1
    Discontinued
    Supplier Number: -
  • 102096-230
    ProScope HR2 CSI Science Level 1
    ProScope HR2 CSI Science Level 1
    Discontinued
    Supplier Number: -
  • 102096-232
    ProScope Wireless CSI Science Level 1
    ProScope Wireless CSI Science Level 1
    Discontinued
    Supplier Number: -
  • 102096-234
    ProScope HR CSI Science Level 2
    ProScope HR CSI Science Level 2
    Discontinued
    Supplier Number: -
  • 102096-236
    ProScope HR2 CSI Science Level 2
    ProScope HR2 CSI Science Level 2
    Discontinued
    Supplier Number: -
  • 102096-238
    ProScope Wireless CSI Science Level 2
    ProScope Wireless CSI Science Level 2
    Discontinued
    Supplier Number: -
  • 102096-256
    ProScope QC Lab
    ProScope QC Lab
    Discontinued
    Supplier Number: 68350-65
  • 102096-258
    ProScope HR2 QC Lab
    ProScope HR2 QC Lab
    Discontinued
    Supplier Number: -
  • 102096-252
    ProScope QC Inspector
    ProScope QC Inspector
    Discontinued
    Supplier Number: -
  • 102096-254
    ProScope HR2 QC Inspector
    ProScope HR2 QC Inspector
    Discontinued
    Supplier Number: -
  • 102096-222
    ProScope HR with 50× Lens
    ProScope HR with 50× Lens
    Log in for availability
    Log in for availability
    Each
    Each - $675.94
    $675.94
    Each
       $675.94
    Supplier Number: 68350-50X
  • 102096-224
    ProScope HR2 with 50× Lens
    ProScope HR2 with 50× Lens
    Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -
Lens
Product Information
  • Availability
    Price
  • 102096-260
    ProScope 0 - 10× Lens
    ProScope 0 - 10× Lens
    Log in for availability
    Log in for availability
    Each
    Each - $288.88
    $288.88
    Each
       $288.88
    Supplier Number: 68352-10
  • 102096-262
    ProScope 100× Lens
    ProScope 100× Lens
    Discontinued
    Supplier Number: -
  • 102096-264
    ProScope 30N Lens
    ProScope 30N Lens
    Discontinued
    Supplier Number: -
  • 102096-266
    ProScope 200× Lens
    ProScope 200× Lens
    Discontinued
    Supplier Number: -
  • 102096-268
    ProScope 400× Lens
    ProScope 400× Lens
    Discontinued
    Supplier Number: -
  • 102096-270
    ProScope 50× Lens
    ProScope 50× Lens
    Discontinued
    Supplier Number: -
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • LX-Proscope HR
    LX-Proscope HR
    102096-298
    Log in for availability
    Each - $307.98
Product Information
  • Catalog #
    Availability
    Price
  • Screen Measurement Pack Mac Edition
    Screen Measurement Pack Mac Edition
    102096-294
    Log in for availability
    Each - $148.62
Product Information
  • Catalog #
    Availability
    Price
  • Screen Calipers for Windows ProScope Edition
    Screen Calipers for Windows ProScope Edition
    102096-292
    Log in for availability
    Each - $58.92
Product Information
  • Catalog #
    Availability
    Price
  • ProScope Deluxe Case
    ProScope Deluxe Case
    102096-284
    Log in for availability
    Each - $152.65
Product Information
  • Catalog #
    Availability
    Price
  • ProScope Case
    ProScope Case
    102096-282
    Log in for availability
    Each - $99.99
Product Information
  • Catalog #
    Availability
    Price
  • ProScope C-Mount Adapter
    ProScope C-Mount Adapter
    102096-272
    Log in for availability
    Each - $46.38
Product Information
  • Catalog #
    Availability
    Price
  • ProScope HR2 Base Unit without Lens
    ProScope HR2 Base Unit without Lens
    102096-216
    Log in for availability
    Each - $465.12
Product Information
  • Catalog #
    Availability
    Price
  • ProScope HR Base Unit without Lens
    ProScope HR Base Unit without Lens
    102096-214
    Log in for availability
    Each - $465.12
Product Information
  • Catalog #
    Availability
    Price
  • ProScope Stand
    ProScope Stand
    102096-278
    Log in for availability
    Each - $307.88
Product Information
  • Catalog #
    Availability
    Price
  • MacCalipers ProScope Edition
    MacCalipers ProScope Edition
    102096-290
    Log in for availability
    Each - $148.62
Product Information
  • Catalog #
    Availability
    Price
  • ProScope Image Analyzer Software
    ProScope Image Analyzer Software
    102096-288
    Log in for availability
    Each - $249.82