Orders LinkContactUsLinkComponent
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Frigimat® Cub dry ice maker
Catalog # 102093-918
Supplier #:  62057-10
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Catalog # 102093-918
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Frigimat® Cub dry ice maker
Price
$1,241.93
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Economical block dry ice maker for on-demand use. In only a few minutes, this compact device can be attached to a liquid CO2 cylinder and will produce a ready-to-use 250 - 300 g block of dry ice.
  • Each 50 lbs. cylinder makes from 10 to 16 blocks of dy ice
  • Set-up and use is safe and simple
  • Dry ice production can be viewed through the transparent polycarbonate body that secures without clamps or screws or tools ensuring easy removal of the solid block of dry ice and connection from the CO2 cylinder
  • A rugged nickel plated “pigtail” connects the Frigimat® Cub – Dry Ice Maker to the CO2 cylinder valve
  • The system operates at very low pressure (displayed on the built-in dial gauge) and incorporates multiple pressure relief safeguards
Cylinder Connection: 1.8 m (6 ft.) nickel-plated copper connecting tube with standard CGA 320 thread coupling for easy attachment to a CO2 cylinder.
Specifications
  • Description:
    Frigimat® Cub dry ice maker