Orders LinkContactUsLinkComponent
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
Certificates
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each complete CoolSafe Deluxe system includes 1 CoolSafe Deluxe box, 1 Cool Chamber, and 2 Cool Bricks (1°C & –10°C).
  • Additional sample protection
  • Smaller bench-top footprint
  • Improved portability
  • Clear lid for sample visibility
CoolSafe for 0.2 mL PCR plates or tubes and the CoolSafe™ Deluxe box also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
Product Family Options
Product Information
  • Availability
    Price
  • 102093-852
    CoolSafe™ Kit for 2 ml tubes or PCR Plates
    CoolSafe™ Kit for 2 ml tubes or PCR Plates
    Log in for availability
    Log in for availability
    Kit of 2
    Kit of 2 - $671.82
    $671.82
    Kit of 2
       $671.82
    Supplier Number: 61954-10
  • 102093-854
    CoolSafe™ Kit for 0.5 ml tubes
    CoolSafe™ Kit for 0.5 ml tubes
    Log in for availability
    Log in for availability
    Kit of 2
    Kit of 2 - $569.20
    $569.20
    Kit of 2
       $569.20
    Supplier Number: 61954-20
  • 102093-858
    CoolSafe™ Kit for flat bottom microplates
    CoolSafe™ Kit for flat bottom microplates
    Log in for availability
    Log in for availability
    Kit of 2
    Kit of 2 - $517.46
    $517.46
    Kit of 2
       $517.46
    Supplier Number: 61954-40