About this item
In high vacuum mode it is ideal for backing turbo pumps, analytical instruments, and electron microscopes. Configurable for high throughput mode. Ultra Grade 19 oil is recommended for use with the RV3.
- Speed - (Pneurop 6602)
- 50 Hz operation - 2 ft 3min-1 (3.3 m3h-1)
- 60 Hz operation - 2.3 ft 3min-1 (3.9 m 3h-1)
- Ultimate Vacuum (Total Pressure) - High Vacuum Mode - 2 x 10-3 mbar
- Inlet connection - NW25