About this item
Low foaming neutral pH detergent for use in manual, and machine cleaning.
- Free rinsing, eliminating interfering residues
- Dilute at 3:100 to get pH 7.0
A safe-handling detergent used for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for aluminum and soft metal, medical devices, pharmaceutical equipments, alkaline sensitive labwares, and medical instruments, aluminum trays.