Orders LinkContactUsLinkComponent
384-Well Plates, Electron Microscopy Sciences
384-Well Plates, Electron Microscopy Sciences
384-Well Plates, Electron Microscopy Sciences
Certificates
384-Well Plates, Electron Microscopy Sciences
384-Well Plates, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Non sterile plates made from natural PP.
Product Family Options
Product Information
  • Well volume
    Availability
    Price
  • 102094-774
    384 Well Plate, Deep Well
    384 Well Plate, Deep Well
    5 - 240 µl
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $1,171.02
    $1,171.02
    Case of 60
       $1,171.02
    Supplier Number: 63498-DW
  • 102094-776
    384 Well Plate, Low Volume
    384 Well Plate, Low Volume
    2 - 35 µl
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $2,206.44
    $2,206.44
    Case of 100
       $2,206.44
    Supplier Number: 63498-LV
  • 102094-778
    384 Well Plate, Standard Volume
    384 Well Plate, Standard Volume
    10 - 100 µl
    Log in for availability
    Log in for availability
    Case of 120
    Case of 120 - $2,051.14
    $2,051.14
    Case of 120
       $2,051.14
    Supplier Number: 63498-ST