Orders LinkContactUsLinkComponent
Poly-L-Lysine Slide, Electron Microscopy Sciences
Poly-L-Lysine Slide, Electron Microscopy Sciences
Poly-l-lysine slide
Catalog # 101412-864
Supplier #:  63478-AS
Poly-L-Lysine Slide, Electron Microscopy Sciences
Catalog # 101412-864
Poly-L-Lysine Slide, Electron Microscopy Sciences
Poly-L-Lysine Slide, Electron Microscopy Sciences
Poly-l-lysine slide
Price
$449.70
Log in for availability
Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Using premium select, low fluorescence glass with a consistent uniform coating of poly-L-lysine these slides yield a dense layer of amine groups for initial ionic attachment of the negatively charged phosphate groups in the DNA backbone.
The DNA can subsequently be attached covalently to the slide by either baking or by UV irradiation. Our aging and packaging techniques ensure a product with a shelf life in excess of 6 months. Each lot is tested for background uniformity prior to shipping. The slides arrive ready for spotting – no pre-washing required. Pre-packaged in a secure, no-contact mBox™, then vacuum-sealed in a foil barrier bag prior to shipping. Each package contains 20 slides.
Specifications
  • Description:
    Poly-l-lysine slide