About this item
Ilford Rapid Fixer is a non-hardening rapid fixer supplied as a liquid concentrate for fixing black and white film and paper in all manual and machine processing applications. It is easy and convenient to use in the temperature range of 18 – 40°C (64 -104°F). Ilford Rapid Fixer must not be used with fix hardeners.