Orders LinkContactUsLinkComponent
Makler Counting Chamber, Electron Microscopy Sciences
Makler Counting Chamber, Electron Microscopy Sciences
Makler Counting Chamber
Catalog # 100499-400
Supplier #:  68052-01
Makler Counting Chamber, Electron Microscopy Sciences
Catalog # 100499-400
Makler Counting Chamber, Electron Microscopy Sciences
Makler Counting Chamber, Electron Microscopy Sciences
Makler Counting Chamber
Price
$3,753.93
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Makler counting chamber is used for rapid semen analysis. It is only 10 microns deep, which is one tenth the depth of other hemacytometers, making it the shallowest of known chambers.
This quality chamber is constructed from two pieces of optically flat glass: the first is the chamber, the second is the cover glass, which has a fine grid pattern of 1mm squares and a center area further subdivided into 0.1 mm squares. Four quartz pins of precise height hold the cover glass to give an exact trapped specimen depth of 10 microns.
Specifications
  • Description:
    Makler Counting Chamber