Flat Embedding Molds, Electron Microscopy Sciences
About this item
Molds are injected molded polyethylene that is resistant to most embedding materials and will easily peeled-off the blocks. These molds are reusable.
- Fits perfectly inside the cocoon box BE8
- Each cavity measures 12L x 5W x 3.5D mm
- Overall mold measures: 94 x 56 x 14 mm
2 Options Available Below
Product Details & Documents
Molds are injected molded polyethylene that is resistant to most embedding materials and will easily peeled-off the blocks. These molds are reusable.
- Fits perfectly inside the cocoon box BE8
- Each cavity measures 12L x 5W x 3.5D mm
- Overall mold measures: 94 x 56 x 14 mm
Product Family Options
Product Information
- AvailabilityPrice
- 102097-196Molded Flat Embedding MoldSupplier Number: 70905-01
- 102097-198Molded Flat Embedding MoldSupplier Number: 70905-12