ContactUsLinkComponent
Incubation Tray for Microwave Immunostaining, Electron Microscopy Sciences
Incubation Tray for Microwave Immunostaining, Electron Microscopy Sciences
  100492-524
Minority or Woman-Owned Business Enterprise
Incubation Tray for Microwave Immunostaining, Electron Microscopy Sciences
  100492-524
 :  97060
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Microwave Incubation Tray for Immunostaining
  • Cat. No.:
    100492-524
  • Supplier no.:
    97060
For immunostaining, we recommend a water load that is the same size as the bottom of the microwave cavity in order not to disturb the distribution of microwave energy within the cavity.
In our research, we found that between 150 and 200 ml of water in the bottom of the incubation tray prevents evaporation of the droplets of reagent. A PAP pen should be used to maintain the droplets at a uniform size and shape.