ContactUsLinkComponent
Horizontal Slide Tray, Electron Microscopy Sciences
Horizontal Slide Tray, Electron Microscopy Sciences
Catalog # 100493-298
Minority or Woman-Owned Business Enterprise
undefined
Horizontal Slide Tray, Electron Microscopy Sciences
Catalog # 100493-298
Supplier Number:  71394-01
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Horizontal Tray
  • Cat. No.:
    100493-298
  • Supplier no.:
    71394-01
Specifications
About this item
This stainless steel tray has grooves to hold various microscope slides horizontally. Accommodates up to 60 slides.