ContactUsLinkComponent
Stainless Steel Biopsy Inserts, Electron Microscopy Sciences
Stainless Steel Biopsy Inserts, Electron Microscopy Sciences
Catalog # 100498-024
Minority or Woman-Owned Business Enterprise
undefined
Stainless Steel Biopsy Inserts, Electron Microscopy Sciences
Catalog # 100498-024
Supplier Number:  62328-01
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Stainless Steel. Biopsy Insert
  • Cat. No.:
    100498-024
  • Fits:
    All process/embedding cassettes and Uni-cassettes
  • Supplier no.:
    62328-01
Specifications
About this item
A two-part loose fitting stainless steel capsule with welded mesh that fits inside the processing cassettes preventing the loss of small or friable material while allowing interchange of fluids.
They fit all process/embedding cassettes such as Process Embedding Cassettes Without Lids, Diamond Dissecting Knife, Type M-M, and Uni-Cassettes.