- Description:Z-6040 Primer
- Size:100 ml
- Cat. No.:100497-424
- Supplier no.:50440-10
A unique embedding primer.
- Better adhesion between resin and specimen
- Prevents sample from pulling away
This unique primer affords you a better adhesion between your resin and specimen that you are embedding and prevents the sample from pulling away from the resin during sectioning.