ContactUsLinkComponent
Vial Holder, Electron Microscopy Sciences
Vial Holder, Electron Microscopy Sciences
Catalog # 100491-986
Minority or Woman-Owned Business Enterprise
undefined
Vial Holder, Electron Microscopy Sciences
Catalog # 100491-986
Supplier Number:  72640
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Shell Vial Holder
  • Cat. No.:
    100491-986
  • Supplier no.:
    72640
Specifications
About this item
Machined from solid aluminum, holds 5 shell vials. It is ideal for handling cold or hot samples for prolonged periods of time out of the refrigerator or oven. Comes complete with a handle. Measurements: 4 3/4 x 1 1/4 x 3/4".