Specifications
- Description:Utility Sheer
- Cat. No.:100502-112
- Supplier no.:72956
Specifications
About this item
Multi purpose sheer with stainless steel blades and plastic handle.
- Autoclavable to 120 °C
One blade is serrated that allows you to cut materials from wire to bone.