Orders LinkContactUsLinkComponent
MAC X-ray Microanalysis Standards Universal Set, Electron Microscopy Sciences
MAC X-ray Microanalysis Standards Universal Set, Electron Microscopy Sciences
MAC X-ray Microanalysis Standards Universal Set, Electron Microscopy Sciences
Certificates
MAC X-ray Microanalysis Standards Universal Set, Electron Microscopy Sciences
MAC X-ray Microanalysis Standards Universal Set, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems
A set of 42 standards for general Energy Dispersive Analysis, mounted in 25 or 32 mm diameter brass stub. Includes Ag, Al2O3, Au, BaF2, Bi, CaSiO3, Cd, CeAl2, Co, Cr, Cu, Fe, FeS2, GaP, Ge, Hf, HgTe, InAs, Ir, KAlSi3O8, LaB6, MgO, Mn, Mo, NaAlSi3O8, Nb, Ni, PbTe, Pd, Pt, Rh, Sb, Se, Sn, SrF2, Ta , Ti, V, W, Y, Zn, and Zr.
All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification.
Product Family Options
Product Information
  • Availability
    Price
  • 100488-086
    Universal Set with 25 mm dia Block
    Universal Set with 25 mm dia Block
    Log in for availability
    Log in for availability
    Kit of 42
    Kit of 42 - $9,179.64
    $9,179.64
    Kit of 42
       $9,179.64
    Supplier Number: 80071-25
  • 100488-088
    Universal Set with 32 mm dia Block
    Universal Set with 32 mm dia Block
    Log in for availability
    Log in for availability
    Kit of 42
    Kit of 42 - $9,179.64
    $9,179.64
    Kit of 42
       $9,179.64
    Supplier Number: 80071-32