Specifications
- Description:Uniband Gripper
- Cat. No.:100492-310
- Supplier no.:76806
Specifications
About this item
This unique Uni-Band gripper is made of 300 Series Memory stainless steel which is harder than titanium. It is perfect gripper for handling specimen mount stubs.