TEM Grid Holder, Electron Microscopy Sciences
Machined from solid steel; has a stainless steel mesh on the top and the bottom.
- Holds 12 grids in numbered slots
Featuring the most advanced design which assures no damage to the grids during processing. These holders are Brass-Nickel plated, very sturdy, and withstand all critical conditions. Dimensions D×H: 24×24 mm.
- Description:Grid holder for CPD