Orders LinkContactUsLinkComponent
PTFE Surface Protectors, Electron Microscopy Sciences
PTFE Surface Protectors, Electron Microscopy Sciences
Tsp, 25"X15ft/Roll
Catalog # 100491-974
Supplier #:  72612
PTFE Surface Protectors, Electron Microscopy Sciences
Catalog # 100491-974
PTFE Surface Protectors, Electron Microscopy Sciences
PTFE Surface Protectors, Electron Microscopy Sciences
Tsp, 25"X15ft/Roll
Price
$615.99
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A thickness of 0.001" "PTFE" film bonded to a 0.008" self-adhesive vinyl gives your work surface good protection.
Easy to use: cut to desired size, remove the backing, and press into place.
Specifications
  • Description:
    Tsp, 25"X15ft/Roll