Orders LinkContactUsLinkComponent
Standard Test Specimen, Electron Microscopy Sciences
Standard Test Specimen, Electron Microscopy Sciences
Standard Test Specimen Set
Catalog # 100502-894
Supplier #:  80154-00
Standard Test Specimen, Electron Microscopy Sciences
Catalog # 100502-894
CAS Number:  
Standard Test Specimen, Electron Microscopy Sciences
Standard Test Specimen, Electron Microscopy Sciences
Standard Test Specimen Set
CAS Number:  
Price
$5,486.68
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The standard test specimen keeps the scanning electron microscope up to peak operating conditions.
  • Brass block of 32 mm diameter consisting of 6 standards
  • Faraday cage
  • Resolution grid either high or medium
  • Test specimen for magnification calibration
There are three formats: Metallurgical, Biological and Geological.
Specifications
Specifications
  • Description:
    Standard Test Specimen Set