ContactUsLinkComponent
Standard Paper Safe, Electron Microscopy Sciences
Standard Paper Safe, Electron Microscopy Sciences
Catalog # 100492-184
Minority or Woman-Owned Business Enterprise
undefined
Standard Paper Safe, Electron Microscopy Sciences
Catalog # 100492-184
Supplier Number:  74377
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Standard Paper Safe
  • Cat. No.:
    100492-184
  • Supplier no.:
    74377
Specifications
About this item
Light proof protection for film and paper. No more fussing with frayed cardboard boxes. Exclusive "Tilt and Tap" feature for easy single sheet removal. Holds 200 Single Weight sheets. Measures 8 1/2 x11" for 8 x 10" paper.