ContactUsLinkComponent
Specimen Gripper, Electron Microscopy Sciences
Specimen Gripper, Electron Microscopy Sciences
Catalog # 100492-030
Minority or Woman-Owned Business Enterprise
undefined
Specimen Gripper, Electron Microscopy Sciences
Catalog # 100492-030
Supplier Number:  72928
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Specimen Gripper
  • Length:
    10¼"
  • Cat. No.:
    100492-030
  • Supplier no.:
    72928
Specifications
About this item
Stainless steel, long, serrated fine tips, for easy removal of specimens from deep containers. Scissor type.