Specifications
- Description:Cassette Storage File Box
- Cat. No.:100500-426
- Supplier no.:63280-20
Specifications
About this item
Moisture resistant, plastic covered corrugated boxes interlock to stack ten high, each supporting up to 500 lbs of overhead cassette and slide weight.
- Economical
- Durable
- Convenient
- Versatile
Each box accommodates 2000 slides or 500 tissue blocks to meet your varied storage requirements. Come unassembled boxes require minimal storage space and can be easily assembled as needed. Slide storage File with four compartments, each has one drawer, drawer inside measures 10½"L x 2 "W x 1½"H (267 x 60 x 40mm), each drawer has one center divider, each division is 11/8" wide, accommodates up to 250 slides or 500 slides per drawer, or 2000 slides per file, 10 stackable files per case or 20,000 slides. When these boxes are used as a cassette storage file, each compartment has two drawers, one on top of the other. Four compartments or eight drawers, each drawer accommodates 63 cassettes or 500 cassettes per file. 10 stackable files per case or 1500 cassettes. Measures 11½" (L) x 12"(W) x 4"(H), (295 x 300 x 100 mm).