ContactUsLinkComponent
Slide Tray Cabinet, Electron Microscopy Sciences
Slide Tray Cabinet, Electron Microscopy Sciences
  100491-788
Minority or Woman-Owned Business Enterprise
undefined
Slide Tray Cabinet, Electron Microscopy Sciences
  100491-788
 :  71504
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Slide Tray Cabinet, 2 × 1 mm Slot
  • Cat. No.:
    100491-788
  • Supplier no.:
    71504
The outside is made of white polypropylene, the tray rails are polystyrene, and the vertical sliding door is clear acrylic; this 240-slide capacity cabinet holds twelve microscope slide trays (#71500) horizontally, and it is dust free. Measurements: 73/4 x 14 x 71/2" high (17 x 36 x 16 cm).